Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP1BB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104021 Supplier Thermo Scientific Supplier No. PA5104021
Add to Cart
Add to Cart

GP1BB Polyclonal Antibody

GP1BB Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GP1BB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GP1BB
Gene Accession No. P13224, P56400
Gene Alias Antigen CD42b-beta; BDPLT1; BS; CD42c; glycoprotein Ib (platelet), beta polypeptide; glycoprotein Ib platelet beta subunit; glycoprotein Ib, beta polypeptide; Gp1bb; GP-Ib beta; GPIbB; GPIbbeta; GPIb-beta; nuclear localization signal deleted in velocardiofacial syndrome; platelet glycoprotein Ib beta chain; platelet membrane glycoprotein Ib beta; truncated platelet membrane glycoprotein Ib beta
Gene Symbols GP1BB
Host Species Rabbit
Immunogen A synthesized peptide derived from human GP1BB(Accession P13224), corresponding to amino acid residues A84-E134.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14724, 2812
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.