Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP1BB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522267 Supplier Invitrogen™ Supplier No. PA522267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji. Predicted reactivity: Mouse (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CD42a is a single-chain membrane glycoprotein that forms a noncovalent complex with CD42b. CD42b, also known as glycoprotein Ib a (GPIb a) is a membrane glycoprotein that is composed of a and b chains. The CD42b b chain is also designated CD42c, and is expressed on platelets and megakaryoctes. CD42a and CD42b are also present on platelets and mega-karyocytes, and the complex is a major component of the platelet surface. The complex acts as a receptor for von Willebrand's factor and as a von Willebrand's factor-dependent adhesion receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP1BB
Applications Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene GP1BB
Gene Accession No. P13224, P56400
Gene Alias Antigen CD42b-beta; BDPLT1; BS; CD42c; glycoprotein Ib (platelet), beta polypeptide; glycoprotein Ib platelet beta subunit; glycoprotein Ib, beta polypeptide; Gp1bb; GP-Ib beta; GPIbB; GPIbbeta; GPIb-beta; nuclear localization signal deleted in velocardiofacial syndrome; platelet glycoprotein Ib beta chain; platelet membrane glycoprotein Ib beta; truncated platelet membrane glycoprotein Ib beta
Gene Symbols GP1BB
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human CD42c. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14724, 2812
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.