Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GP2 Antibody (GP2/1805), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308215AF594
Description
GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
GP2 | |
Monoclonal | |
Alexa Fluor 594 | |
DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary) (Uniprot: P55259) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array | |
GP2/1805 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
2813 | |
Store at 4°C in the dark. | |
IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction