Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GP2 Antibody (GP2/3134R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308243
Description
GP2 Monoclonal specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
GP2 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, ELISA : 2-4 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL | |
DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
GP2/3134R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
2813 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction