Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP2 Mouse anti-Human, Clone: GP2/1805, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen GP2
Clone GP2/1805
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122128
SDP
View Documents
Novus Biologicals
NBP30732720UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

GP2 Monoclonal specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

GP2
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Human
DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75
Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary)
Primary
Store at 4°C.
GP2/1805
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
2813
IgG2c κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.