Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592932 Supplier Invitrogen™ Supplier No. PA592932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSSILQTEER NWVSVTSPVQ ASACRNILER NQTHAIYKNT LSLVNDFIIR DTILNINFQC AYPLDMKVSL QAALQPIVSS LNVSVDGNGE FIVRMALFQD QNYTNPYEGD AVELSVESVL YVGAILEQGD TSRFNLVLRN CYATPTEDKA DLVKYFIIRN SCSNQRDSTI HVEENGQSSE SRFSVQMFMF AGHYDLVFLH CEIHLCDSLN EQCQPSCSRS QVRSEVPAID LARVLDLGPI TRRGAQSPGV MNGTPSTAGF LVAWPMVLLT VLLAWLF; Positive Samples: U-251MG; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.

GP2 is component of pancreatic secretory (zymogen) granule. The exact function of GP2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GP2
Gene Accession No. P55259
Gene Alias 2310037I18Rik; AV060639; glycoprotein 2; glycoprotein 2 (zymogen granule membrane); glycoprotein 80; GP2; gp80; Pancreatic secretory granule membrane major glycoprotein GP2; pancreatic zymogen granule membrane associated protein GP2; pancreatic zymogen granule membrane protein GP-2; secretory (zymogen) granule membrane glycoprotein GP2; ZAP75; zymogen granule membrane glycoprotein 2
Gene Symbols GP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 111-387 of human GP2 (NP_0010072432).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2813
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.