Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP2 Rabbit anti-Human, Unconjugated, Clone: GP2/3134R, Abnova™

Catalog No. 89337846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-846 Supplier Abnova Corporation Supplier No. RAB00421
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for ELISA, IHC-P.

Specifications

Antigen GP2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GP2/3134R
Conjugate Unconjugated
Description glycoprotein 2 (zymogen granule membrane)
Gene GP2
Gene Alias DKFZp779K0533, ZAP75
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to amino acid 35-179 of human GP2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2813
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.