Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gp210 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129888
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129888 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129888 Supplier Invitrogen™ Supplier No. PA129888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29888 detects GP210 from mouse, non-human primate, human, rat samples.

NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gp210
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.1% sodium azide
Gene NUP210
Gene Accession No. P11654, Q8TEM1, Q9QY81
Gene Alias 9830001L10; AI836801; FLJ22389; gp190; GP210; integral membrane nucleoporin gp210; Kiaa0906; Nuclear envelope pore membrane protein POM 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 210kDa L homeolog; nucleoporin Nup210; NUP210; nup210.L; POM210; pore membrane protein of 210 kDa; PSEC0245; XELAEV_18001839mg
Gene Symbols NUP210
Host Species Rabbit
Immunogen Purified bacterial C-terminal fusion protein.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23225, 54563, 58958
Target Species Human, Mouse, Monkey, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.