Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gp210 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111617 Supplier Invitrogen™ Supplier No. PA5111617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TFTVHFHDNS GDVFHAHSSV LNFATNRDDF VQIGKGPTNN TCVVRTVSVG LTLLRVWDAE HPGLSDFMPL PVLQAISPEL SGAMVVGDVL CLATVLTSLE GLSGTWSSSA NSILHIDPKT GVAV Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 81%.

NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gp210
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUP210
Gene Accession No. Q8TEM1
Gene Alias 9830001L10; AI836801; FLJ22389; gp190; GP210; integral membrane nucleoporin gp210; Kiaa0906; Nuclear envelope pore membrane protein POM 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 210kDa L homeolog; nucleoporin Nup210; NUP210; nup210.L; POM210; pore membrane protein of 210 kDa; PSEC0245; XELAEV_18001839mg
Gene Symbols NUP210
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human gp210. Recombinant protein control fragment (Product #RP-103643).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23225
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.