Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ gp210 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115312
Description
Antibody detects endogenous levels of total GP210.
NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
Specifications
gp210 | |
Polyclonal | |
Unconjugated | |
NUP210 | |
9830001L10; AI836801; FLJ22389; gp190; GP210; integral membrane nucleoporin gp210; Kiaa0906; Nuclear envelope pore membrane protein POM 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 210kDa L homeolog; nucleoporin Nup210; NUP210; nup210.L; POM210; pore membrane protein of 210 kDa; PSEC0245; XELAEV_18001839mg | |
Rabbit | |
Affinity chromatography | |
RUO | |
23225, 54563, 58958 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P11654, Q8TEM1, Q9QY81 | |
NUP210 | |
A synthesized peptide derived from human NUP210(Accession Q8TEM1), corresponding to amino acid residues H1851-H1887. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction