Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP340 Monoclonal Antibody (548031)
GREENER_CHOICE

Catalog No. PIMA524249
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524249 Supplier Invitrogen™ Supplier No. MA524249
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) CD163 and rhSRCRB4D is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Glycoprotein-340 is member of the scavenger receptor superfamily and has been identified as a putative SP-D receptor. Gp-340 has been shown to bind SP-D in a calcium dependent manner through a protein-protein interaction. Like SP-D, gp-340 is mainly found in lung tissue and immunohistochemistry revealed high expression in alveolar macrophages. An identical protein is present in saliva (salivary agglutinin) and both forms are encoded by the gene DMBT1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP340
Applications Flow Cytometry
Classification Monoclonal
Clone 548031
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene DMBT1
Gene Accession No. Q60997
Gene Alias apactin; CRP; CRP-[a]; CRP-[b]; Crpd; CRP-ductin; DBMT1; deleted in malignant brain tumors 1; deleted in malignant brain tumors 1 protein; deleted in malignant brain tumors 1 pseudogene; DMBT1; Ebnerin; glycoprotein 300; Glycoprotein 340; gp300; GP340; gp-340; hensin; Mucin-like glycoprotein; muclin; p80; pancrin; RP11-481L19.1; SAG; Salivary agglutinin; surfactant pulmonary-associated D-binding protein; Vomeroglandin
Gene Symbols DMBT1
Host Species Rat
Immunogen E. Coli-derived recombinant mouse DMBT1 Trp215-Gly420.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12945
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.