Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP340 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107112 Supplier Invitrogen™ Supplier No. PA5107112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GP340 Polyclonal Antibody for SPR, Western Blot

Glycoprotein-340 is member of the scavenger receptor superfamily and has been identified as a putative SP-D receptor. Gp-340 has been shown to bind SP-D in a calcium dependent manner through a protein-protein interaction. Like SP-D, gp-340 is mainly found in lung tissue and immunohistochemistry revealed high expression in alveolar macrophages. An identical protein is present in saliva (salivary agglutinin) and both forms are encoded by the gene DMBT1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP340
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene DMBT1
Gene Accession No. Q9UGM3
Gene Alias apactin; CRP; CRP-[a]; CRP-[b]; Crpd; CRP-ductin; DBMT1; deleted in malignant brain tumors 1; deleted in malignant brain tumors 1 protein; deleted in malignant brain tumors 1 pseudogene; DMBT1; Ebnerin; glycoprotein 300; Glycoprotein 340; gp300; GP340; gp-340; hensin; Mucin-like glycoprotein; muclin; p80; pancrin; RP11-481L19.1; SAG; Salivary agglutinin; surfactant pulmonary-associated D-binding protein; Vomeroglandin
Gene Symbols DMBT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human DMBT1 (aa 467-516).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.