Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144819
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144819 Supplier Invitrogen™ Supplier No. PA5144819
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADPEIKRGSG WRPTGCSQPR VMFMTAEPQA RSYPREGSWH GRRLKDWRVW SVEAGGQRLQ LWKRGHAASS WCSIREPFGQ CLSVCLP Antigen sequence identity: Mouse 25%; Rat 22%.

Glycoprotein VI (GP6) is a 58-kD platelet membrane glycoprotein that plays a crucial role in the collagen-induced activation and aggregation of platelets. Upon injury to the vessel wall and subsequent damage to the endothelial lining, exposure of the subendothelial matrix to blood flow results in deposition of platelets. Collagen fibers are the most thrombogenic macromolecular components of the extracellular matrix, with collagen types I, III, and VI being the major forms found in blood vessels. Platelet interaction with collagen occurs as a 2-step procedure: (1) the initial adhesion to collagen is followed by (2) an activation step leading to platelet secretion, recruitment of additional platelets, and aggregation. In physiologic conditions, the resulting platelet plug is the initial hemostatic event limiting blood loss. However, exposure of collagen after rupture of atherosclerotic plaques is a major stimulus of thrombus formation associated with myocardial infarction or stroke.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GP6
Gene Accession No. Q9HCN6
Gene Alias 9830166G18Rik; BDPLT11; Glycoprotein 5; glycoprotein 6; glycoprotein 6 (platelet); glycoprotein VI (platelet); glycoprotein VI platelet; Gm469; GP6; GPIV; GPVI; platelet collagen receptor; platelet glycoprotein VI
Gene Symbols GP6
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 318-404. Recombinant protein control fragment (Product #RP-110189).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51206
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.