Learn More
Invitrogen™ GP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590172
Description
Positive Samples: Jurkat, human plasma; Cellular Location: Cell membrane, Single-pass membrane protein Immunogen sequence: QSGPLPKPSL QALPSSLVPL EKPVTLRCQG PPGVDLYRLE KLSSSRYQDQ AVLFIPAMKR SLAGRYRCSY QNGSLWSLPS DQLELVATGV FAKPSLSAQP GPAVSSGGDV TLQCQTRYGF DQFALYKEGD PAPYKNPERW YRASFPIITV TAAHSGTYRC YSFSSRDPYL WSAPSDPLEL VVTGTSVTPS RLPTEPPSPV AEFSEATAEL TVSFTNEVFT TETSRSITAS PKESDSPAGP ARQYYTK.
Glycoprotein VI (GP6) is a 58-kD platelet membrane glycoprotein that plays a crucial role in the collagen-induced activation and aggregation of platelets. Upon injury to the vessel wall and subsequent damage to the endothelial lining, exposure of the subendothelial matrix to blood flow results in deposition of platelets. Collagen fibers are the most thrombogenic macromolecular components of the extracellular matrix, with collagen types I, III, and VI being the major forms found in blood vessels. Platelet interaction with collagen occurs as a 2-step procedure: (1) the initial adhesion to collagen is followed by (2) an activation step leading to platelet secretion, recruitment of additional platelets, and aggregation. In physiologic conditions, the resulting platelet plug is the initial hemostatic event limiting blood loss. However, exposure of collagen after rupture of atherosclerotic plaques is a major stimulus of thrombus formation associated with myocardial infarction or stroke.
Specifications
GP6 | |
Polyclonal | |
Unconjugated | |
GP6 | |
9830166G18Rik; BDPLT11; Glycoprotein 5; glycoprotein 6; glycoprotein 6 (platelet); glycoprotein VI (platelet); glycoprotein VI platelet; Gm469; GP6; GPIV; GPVI; platelet collagen receptor; platelet glycoprotein VI | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51206 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9HCN6 | |
GP6 | |
A synthetic peptide corresponding to a sequence within amino acids 21-267 of human GP6 (NP_0574475). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.