Learn More
Invitrogen™ GP6 Polyclonal Antibody

Description
A suggested positive control is A-20 cell lysate. PA5-20583 can be used with blocking peptide PEP-0703.
Specifications
Specifications
| Antigen | GP6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GP6 |
| Gene Accession No. | P0C191, Q9HCN6 |
| Gene Alias | 9830166G18Rik; BDPLT11; Glycoprotein 5; glycoprotein 6; glycoprotein 6 (platelet); glycoprotein VI (platelet); glycoprotein VI platelet; Gm469; GP6; GPIV; GPVI; platelet collagen receptor; platelet glycoprotein VI |
| Gene Symbols | GP6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.