Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gp96/HSP90B1/GRP94 Antibody (HSP90B1/1192), Janelia Fluor™ 549, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP311548JF549
Description
gp96/HSP90B1/GRP94 Monoclonal antibody specifically detects gp96/HSP90B1/GRP94 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
gp96/HSP90B1/GRP94 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HSP90B1/1192 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96) | |
Recombinant full-length human gp96/HSP90B1/GRP94 protein (Uniprot: P14625) | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cellular Markers, Cytokine Research, ER Markers, Golgi Apparatus Markers, Hypoxia, Immunology, Membrane Trafficking and Chaperones, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
7184 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction