Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gp96/HSP90B1/GRP94 Antibody (SPM249) - IHC-Prediluted, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP248051
Description
gp96/HSP90B1/GRP94 Monoclonal specifically detects gp96/HSP90B1/GRP94 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine, Guinea Pig, Hamster, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
gp96/HSP90B1/GRP94 | |
Monoclonal | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
HSP90B1 | |
Purified glucose regulated protein 94 (grp94) from chicken oviducts (Uniprot: P14625) | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 94kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This monoclonal antibody is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grps are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer. | |
Store at 4C. | |
IgG2a κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM249 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96) | |
Rat | |
94 kDa | |
7 mL | |
Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
7184 | |
Human, Mouse, Rat, Pig, Canine, Chicken, Equine, Guinea Pig, Hamster, Primate, Monkey, Rabbit, Sheep, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction