Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPALPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566199 Supplier Invitrogen™ Supplier No. PA566199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGPALPPGFKK QDDSPPRPII GPALPPGFIK STQKSDKGRD DPGQQETDSS EDEDIIGPMP AKGPVNYNVT TEFEKRAQRM KE Highest antigen sequence identity to the following orthologs - mouse 82%, rat 79%.

GPALPP1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPALPP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPALPP1
Gene Accession No. Q8IXQ4
Gene Alias 1200011I18Rik; AD029; bA245H20.2; C6orf72; dJ12G14.2; GINM1; glycoprotein integral membrane 1; glycoprotein integral membrane protein 1; GPALPP motifs containing 1; GPALPP motifs-containing protein 1; Gpalpp1; hypothetical protein LOC532698; hypothetical protein LOC55425; Kiaa1704; lipopolysaccharide specific response-7 protein; lipopolysaccharide-specific response protein 7; LSR7; RGD1307254; UNQ710/PRO1361
Gene Symbols GPALPP1
Host Species Rabbit
Immunogen Recombinant Human GPALPP1. Recombinant protein control fragment (Product #RP-105109).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55425
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.