Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPALPP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570300
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
GPALPP1 is a protein coding gene.
Specifications
GPALPP1 | |
Polyclonal | |
Unconjugated | |
GPALPP1 | |
1200011I18Rik; AD029; bA245H20.2; C6orf72; dJ12G14.2; GINM1; glycoprotein integral membrane 1; glycoprotein integral membrane protein 1; GPALPP motifs containing 1; GPALPP motifs-containing protein 1; Gpalpp1; hypothetical protein LOC532698; hypothetical protein LOC55425; Kiaa1704; lipopolysaccharide specific response-7 protein; lipopolysaccharide-specific response protein 7; LSR7; RGD1307254; UNQ710/PRO1361 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55425 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IXQ4 | |
GPALPP1 | |
synthetic peptide directed towards the middle region of human GPALPP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction