Learn More
Invitrogen™ GPAM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593092
Description
Immunogen sequence: LNKRGLGGPT STPPNLISQE QLVRKAASLC YLLSNEGTIS LPCQTFYQVC HETVGKFIQY GILTVAEHDD QEDISPSLAE QQWDKKLPEP LSWRSDEEDE DSDFGEEQRD CYLKVSQSKE HQQFITFLQR LLGPLLEAYS SAAIFVHNFS GPVPEPEYLQ KLHKYLITRT ERNVAVYAES ATYCLVKNAV KMFKDIGVFK ETKQKRVSVL ELSSTFLPQC NRQKLLEYIL SFVVL; Positive Samples: Mouse heart, Mouse lung, Rat brain; Cellular Location: Mitochondrion outer membrane, Multi-pass membrane protein.
Glycerol-3-phosphate acyltransferase 1 (GPAT1), one of four known GPAT isoforms, is located on the mitochondrial outer membrane, allowing reciprocal regulation with carnitine palmitoyltransferase-1. It is thought to be critical for the development of hepatic steatosis; steatosis triggered by GPAT1 overexpression leads to hepatic and possibly peripheral insulin resistance. GPAT1 is transcriptionally upregulated by insulin and sterol regulatory element binding protein (SREBP-1) and downregulated by AMP-activated protein kinase. Mice deficient in GPAT1 exhibit decreased triacylglycerol (TAG) in cardiomyocytes even in high-fat diets, suggesting that GPAT1 contributes significantly to TAG accumulation in heart tissue during lipogenic or high fat diets. At least two isoforms of GPAT1 are known to exist.
Specifications
GPAM | |
Polyclonal | |
Unconjugated | |
Gpam | |
glycerol 3-phosphate acyltransferase, mitochondrial; glycerol-3-phosphate acyltransferase 1, mitochondrial; glycerol-3-phosphate acyltransferase, mitochondrial; GPAM; GPAT; GPAT1; GPAT-1; KIAA1560; P90; RP11-426E5.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14732, 29653, 57678 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97564, Q61586, Q9HCL2 | |
Gpam | |
Recombinant fusion protein containing a sequence corresponding to amino acids 594-828 of human GPAM (NP_0012318781). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.