Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPANK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552275 Supplier Invitrogen™ Supplier No. PA552275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVCELSGRDA AQLAEEAGFP EVARMVRESH GETRSPENRS PTPSLQYCEN CDTHFQDSNH RTSTAHLLSL SQGPQPPNLP LGVPISSPGF KLLLRGGWEP GMGLGPRGEG RANPIPTVLK RDQEGLGYRS APQ Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

This gene is located in a cluster of HLA-B-associated transcripts, which is included in the human major histocompatability complex III region. This gene encodes a protein which is thought to play a role in immunity. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Nov 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPANK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPANK1
Gene Accession No. O95872
Gene Alias ANKRD59; Ankyrin repeat domain-containing protein 59; BAT4; Bat-4; D17H6S54E; D6S54E; G patch domain and ankyrin repeat-containing protein 1; G patch domain and ankyrin repeats 1; G patch domain and ankyrin repeats-containing protein 1; G patch domain containing 10; g patch domain-containing protein 10; G5; G5 protein; Gpank1; G-patch domain and ankyrin repeats 1; GPATCH10; HLA-B associated transcript 4; HLA-B-associated transcript 4; protein BAT4; protein G5
Gene Symbols GPANK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPANK1. Recombinant protein control fragment (Product #RP-102481).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7918
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.