Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115315
Description
Antibody detects endogenous levels of total GPAT-2.
GPAT2 gene ontology annotations related to this gene include cellular lipid metabolic process.
Specifications
GPAT2 | |
Polyclonal | |
Unconjugated | |
GPAT2 | |
A530057A03Rik; cancer/testis antigen 123; CT123; Glycerol-3-phosphate acyltransferase 2, mitochondrial; glycerol-3-phosphate acyltransferase-like 1; Gm116; GPAT2; GPAT-2; RGD1304904; xGPAT1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
150763, 215456, 296130 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q14DK4, Q6NUI2 | |
GPAT2 | |
A synthesized peptide derived from human GPAT2(Accession Q6NUI2), corresponding to amino acid residues C615-D665. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction