Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPAT4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15587R

Description
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.Specifications
GPAT4 | |
Polyclonal | |
Unconjugated | |
GPAT4 | |
1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-AGP; 1-AGP acyltransferase 6; 1-AGPAT 6; Acyl-CoA:glycerol-3-phosphate acyltransferase 4; Agpat6; AU041707; AW545732; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; Gpat4; LPAATZ; LPAAT-zeta; Lysophosphatidic acid acyltransferase zeta; putative lysophosphatidic acid acyltransferase; RGD1310520; Testis spermatogenesis apoptosis-related protein 7; Tsarg7; UNQ551/PRO1108 | |
Rabbit | |
Protein A | |
RUO | |
137964 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q86UL3 | |
GPAT4 | |
KLH conjugated synthetic peptide derived from human GPAT4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction