Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPATCH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145069
Description
Immunogen sequence: RYVGKILDGF SLASKPLSSK KIYPPPELPR DYRPVHYFRP MVAATSENSH LLQVLSESAG KATPDPGTHS KHQLNASKRA ELLGETPIQG SATSVL Antigen sequence identity: Mouse 86%; Rat 88%.

Specifications
GPATCH1 | |
Polyclonal | |
Unconjugated | |
GPATCH1 | |
1300003A17Rik; ECGP; evolutionarily conserved G patch domain containing; evolutionarily conserved G-patch domain containing; evolutionarily conserved G-patch domain-containing protein; G patch domain containing 1; G patch domain-containing protein 1; GPATC1; G-patch domain containing 1; Gpatch1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55094 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRR8 | |
GPATCH1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 340-435. Recombinant protein control fragment (Product #RP-109755). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction