Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPATCH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555584 Supplier Invitrogen™ Supplier No. PA555584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHPWETGHCL SEGSDSSLEE PSKDYRENHN NNKKDHSDSD DQMLVAKRRP SSNLNNNVRG KRPLWHESDF Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

The gene encodes a nuclear factor that may play a role in spermatogenesis and in tumor growth during breast cancer. The encoded protein contains a G-patch domain with an RNA binding motif. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPATCH2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPATCH2
Gene Accession No. Q9NW75
Gene Alias 5830433G22Rik; 5830436K05Rik; AI427714; AI447508; AW107440; AW491060; cancer/testis antigen 110; CT110; G patch domain containing 2; G patch domain-containing protein 2; GPATC2; G-patch domain containing 2; GPATCH2; PPP1R30; protein phosphatase 1, regulatory subunit 30; RP11-361K17.1; si:ch211-57i17.3; si:ch73-21k16.4
Gene Symbols GPATCH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPATCH2. Recombinant protein control fragment (Product #RP-94052).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55105
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.