Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPATCH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570280
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
The gene encodes a nuclear factor that may play a role in spermatogenesis and in tumor growth during breast cancer. The encoded protein contains a G-patch domain with an RNA binding motif. Alternative splicing results in multiple transcript variants.
Specifications
GPATCH2 | |
Polyclonal | |
Unconjugated | |
GPATCH2 | |
5830433G22Rik; 5830436K05Rik; AI427714; AI447508; AW107440; AW491060; cancer/testis antigen 110; CT110; G patch domain containing 2; G patch domain-containing protein 2; GPATC2; G-patch domain containing 2; GPATCH2; PPP1R30; protein phosphatase 1, regulatory subunit 30; RP11-361K17.1; si:ch211-57i17.3; si:ch73-21k16.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
289362 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
GPATCH2 | |
synthetic peptide directed towards the C-terminal region of rat GPATCH2. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction