Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPATCH2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982921
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2921 Supplier Bioss Supplier No. BS13499R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The gene encodes a nuclear factor that may play a role in spermatogenesis and in tumor growth during breast cancer. The encoded protein contains a G-patch domain with an RNA binding motif. Alternative splicing results in multiple transcript variants.

Specifications

Antigen GPATCH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPATCH2
Gene Accession No. Q9NW75
Gene Alias 5830433G22Rik; 5830436K05Rik; AI427714; AI447508; AW107440; AW491060; BOS_15633; cancer/testis antigen 110; CT110; G patch domain containing 2; G patch domain-containing protein 2; GPATC2; G-patch domain containing 2; Gpatch2; PPP1R30; protein phosphatase 1, regulatory subunit 30; RP11-361K17.1; si:ch211-57i17.3; si:ch73-21k16.4
Gene Symbols GPATCH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPATCH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55105
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.