Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPATCH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565909 Supplier Invitrogen™ Supplier No. PA565909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHTKGIGRKVM ERQGWAEGQG LGCRCSGVPE ALDSDGQHPR CKRGLGYHGE KLQPFGQLKR PRRNGLGLIS TIYDEPLPQD QTES Highest antigen sequence identity to the following orthologs - mouse 93%, rat 89%.

GPATCH3 is a protein coding gene involved in transcriptional regulation. It is able to activate transcription from the CXCR4 promoter and therefore it might control neural crest cell migration involved in ocular and craniofacial development. Is a negative regulator of immune antiviral response, acting via down-regulation of RIG-I-like receptors signaling and inhibition of type I interferon production. The control mechanism involves interaction with mitochondrial MAVS and inhibition of MAVS assembly with downstream proteins implicated in antiviral response, such as TBK1 and TRAF6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPATCH3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPATCH3
Gene Accession No. Q96I76
Gene Alias D930035B09Rik; G patch domain containing 3; G patch domain-containing protein 3; Gpatc3; G-patch domain containing 3; GPATCH3; RGD1311216
Gene Symbols GPATCH3
Host Species Rabbit
Immunogen Recombinant Human GPATCH3. Recombinant protein control fragment (Product #RP-105006).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.