Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPATCH3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982922
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2922 Supplier Bioss Supplier No. BS13500R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPATCH3 is a protein coding gene involved in transcriptional regulation. It is able to activate transcription from the CXCR4 promoter and therefore it might control neural crest cell migration involved in ocular and craniofacial development. Is a negative regulator of immune antiviral response, acting via down-regulation of RIG-I-like receptors signaling and inhibition of type I interferon production. The control mechanism involves interaction with mitochondrial MAVS and inhibition of MAVS assembly with downstream proteins implicated in antiviral response, such as TBK1 and TRAF6.

Specifications

Antigen GPATCH3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPATCH3
Gene Accession No. Q96I76
Gene Alias G patch domain containing 3; G patch domain-containing protein 3; GPATC3; G-patch domain containing 3; GPATCH3
Gene Symbols GPATCH3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPATCH3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63906
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.