Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPATCH4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545396
Description
Peptide sequence: KKRRHQEGKV SDEREGTTKG NEKEDAAGTS GLGELNSREQ TNQSLRKGKK Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
GPATCH4 | |
Polyclonal | |
Unconjugated | |
GPATCH4 | |
2610029K21Rik; DKFZP434F1735; FLJ20249; G patch domain containing 4; G patch domain-containing protein 4; Gpatc4; G-patch domain containing 4; Gpatch4; RGD1311086 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54865 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T3I0 | |
GPATCH4 | |
Synthetic peptide directed towards the C-terminal of human GPATCH4 (aa 243-292). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction