Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPATCH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565858 Supplier Invitrogen™ Supplier No. PA565858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGMKFAEEQLL KHGWTQGKGL GRKENGITQA LRVTLKQDTH GVGHDPAKEF TNHWWNELFN KTAANLVVET GQDGVQIRSL SKETTRY Highest antigen sequence identity to the following orthologs - mouse 85%, rat 89%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPATCH4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPATCH4
Gene Accession No. Q5T3I0
Gene Alias 2610029K21Rik; DKFZP434F1735; FLJ20249; G patch domain containing 4; G patch domain-containing protein 4; Gpatc4; G-patch domain containing 4; Gpatch4; RGD1311086
Gene Symbols GPATCH4
Host Species Rabbit
Immunogen Recombinant Human GPATCH4. Recombinant protein control fragment (Product #RP-104995).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54865
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.