Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPBB Monoclonal Antibody (6F1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA121530
Description
MA1-21530 detects GPBB in Human samples.
The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
Specifications
GPBB | |
Monoclonal | |
2.0 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P11216 | |
PYGB | |
Recombinant full length protein (Human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Affinity Purification, ELISA, Western Blot | |
6F1 | |
Unconjugated | |
PYGB | |
brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB | |
Mouse | |
Protein A | |
RUO | |
5834 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction