Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPBB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583180 Supplier Invitrogen™ Supplier No. PA583180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTMDGANVEM AEEAGAENLF IFGLRVEDVE ALDRKGYNAR EYYDHLPELK QAVDQISSGF FSPKEPDCFK DIVNMLMHHD RFKVFADYEA YMQCQAQVDQ LYRNPKEWTK KVIRNIACSG KFSSDRTITE YAREIWGVEP SDLQIPP Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 91%.

The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPBB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PYGB
Gene Accession No. P11216
Gene Alias brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB
Gene Symbols PYGB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PYGB. Recombinant protein control fragment (Product #RP-102391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.