Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPBB Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589290 Supplier Thermo Scientific Supplier No. PA589290

Rabbit Polyclonal Antibody

Immunogen sequence: LKFSAFLEKE YKVKINPSSM FDVHVKRIHE YKRQLLNCLH VVTLYNRIKR DPAKAFVPRT VMIGGKAAPG YHMAKLIIKL VTSIGDVVNH DPVVGDRLKV IFLENYRVSL AEKVIPAADL SQQISTAGTE ASGTGNMKFM LNGALTIGTM DGANVEMAEE AGAENLFIFG LRVEDVEALD RKGYNAREYY DHLPELKQAV DQISSGFFSP KEPDCFKDIV NMLMHHDRFK VFADYEAYMQ CQAQVDQLYR NPKEWTKKVI RNIACSGKFS SDRTITEYAR EIWGVEPSDL QIPPPNIPRD; Positive Samples: BT-474, SW620, HepG2, Mouse lung, Rat brain

The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPBB
Applications Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene PYGB
Gene Accession No. P11216, P53534, Q8CI94
Gene Alias brain glycogen phosphorylase, glycogen phosphorylase B, glycogen phosphorylase, brain form, GLYPHOA, GPBB, MGC9213, phosphorylase, glycogen, brain, PYGB
Gene Symbols PYGB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 544-843 of human PYGB (NP_0028532)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110078, 25739, 5834
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.