Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPBP1/Vasculin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982924
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2924 Supplier Bioss Supplier No. BS13503R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vasculin is a novel vascular protein differentially expressed in human atherogenesis.

Specifications

Antigen GPBP1/Vasculin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPBP1
Gene Accession No. Q86WP2
Gene Alias 1700034P14Rik; AU019836; BOS_19737; D230035M11Rik; DKFZP761C169; GC-rich promoter binding protein 1; GC-rich promoter-binding protein 1; Gpbp; GPBP1; LOW QUALITY PROTEIN: vasculin; mGPBP; RGD1305492; SSH6; Vascular wall-linked protein; Vasculin
Gene Symbols GPBP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPBP1/Vasculin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65056
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.