Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPBP1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578552 Supplier Invitrogen™ Supplier No. PA578552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, HeLa, HepG2 Predicted Reactivity: Mouse (88%), Rat (85%), Chicken (80%), Rhesus Monkey (98%), Bovine (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GPBP1L1 is a possible transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPBP1L1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Gpbp1l1
Gene Accession No. Q9HC44
Gene Alias 5330440M15Rik; BC002292; GC-rich promoter binding protein 1 like 1; GC-rich promoter binding protein 1-like 1; GC-rich promoter-binding protein 1-like 1; GPBP1L1; RP11-767N6.1; SP192; vasculin-like protein 1
Gene Symbols Gpbp1l1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GPBP1L1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.