Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPC3 Mouse anti-Human, Unconjugated, Clone: 1G12 + GPC3/863, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB21015
Description
GPC3 Monoclonal antibody specifically detects GPC3 in Human samples. It is validated for IHC-P, IF, Flow Cyt.Specifications
GPC3 | |
Monoclonal | |
Unconjugated | |
GPC3 | |
Mouse | |
100 μg | |
2719 | |
Store at 2 to 8°C | |
IgG |
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry | |
1G12 + GPC3/863 | |
glypican 3 | |
DGSX, OCI-5, SDYS, SGB, SGBS, SGBS1 | |
Recombinant protein corresponding to amino acid 511-580 of human GPC3 (1G12); Recombinant protein corresponding to full length human GPC3 (GPC3/863) | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction