Learn More
GPC3 Rabbit anti-Human, Rat, Unconjugated, Clone: GPC3/1534R, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | GPC3 |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Clone | GPC3/1534R |
| Conjugate | Unconjugated |
| Description | glypican 3 |
| Gene | GPC3 |
| Gene Alias | DGSX, OCI-5, SDYS, SGB, SGBS, SGBS1 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein corresponding to full-length human GPC3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.