Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPC3 Rabbit anti-Human, Rat, Unconjugated, Clone: GPC3/1534R, Abnova™

Catalog No. 89339744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-744 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-744 Supplier Abnova Corporation Supplier No. RAB00705
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

GPC3 Monoclonal antibody specifically detects GPC3 in Human, Rat samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen GPC3
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone GPC3/1534R
Conjugate Unconjugated
Description glypican 3
Gene GPC3
Gene Alias DGSX, OCI-5, SDYS, SGB, SGBS, SGBS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human GPC3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2719
Target Species Human, Rat
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.