Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPCPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565346 Supplier Invitrogen™ Supplier No. PA565346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAVALLPENDT GESMLWKATI VLSRGVSVQY RYFKGYFLEP KTIGGPCQVI VHKWETHLQP RSITPLESEI IIDDGQFGIH NGVETLDSGW LTCQTEIRLR LHYSEKPPVS ITKKKLKKSR FRVKLTLEGL EEDDDDRVSP TVLHKMS Highest antigen sequence identity to the following orthologs - mouse 88%, rat 87%.

May be involved in the negative regulation of skeletal muscle differentiation, independently of its glycerophosphocholine phosphodiesterase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPCPD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPCPD1
Gene Accession No. Q9NPB8
Gene Alias EDI3; endometrial differential 3; GDE5; GDPD6; glycerophosphocholine phosphodiesterase 1; glycerophosphocholine phosphodiesterase GDE1 homolog; glycerophosphocholine phosphodiesterase GPCPD1; Glycerophosphodiester phosphodiesterase 5; GPCPD1; KIAA1434; PREI4; preimplantation protein 4; putative glycerophosphocholine phosphodiesterase GPCPD1; putative glycerophosphodiester phosphodiesterase 5
Gene Symbols GPCPD1
Host Species Rabbit
Immunogen Recombinant Human GPCPD1. Recombinant protein control fragment (Product #RP-106035).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56261
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.