Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPCR GPR3 Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207240
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7240 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7240 Supplier Fabgennix Inc Supplier No. GPCR3101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPCR GPR3 Polyclonal antibody specifically detects GPCR GPR3 in Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen GPCR GPR3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene GPR3
Gene Accession No. P35413, Q8K1Q3
Gene Alias ACCA; ACCA orphan receptor; adenylate cyclase constitutive activator; G protein-coupled receptor 3; G protein-coupled receptor R4; Gpcr20; Gpcr21; Gpcr3; GPR3; G-protein coupled receptor 3; G-protein-coupled receptor R4; Hacca; OTTHUMP00000043209
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 5-50 on rat GPCR3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14748, 266769
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.