Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGD Polyclonal Antibody

GPCR LS195564 Polyclonal Antibody

Supplier:  Invitrogen™ PA132863

Catalog No. PIPA132863


Only null left
Add to Cart

Description

Description

PA1-32863 detects GPCR LS195564 from human samples.

The G protein-coupled receptor LS195564 is an Orphan-A GPCR with an unknown ligand.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPCR LS195564
Polyclonal
Unconjugated
MRGPRD
MRGPRD
Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human TGR7 / MRGD.
25 μg
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q8TDS7
Rabbit
Antigen affinity chromatography
RUO
116512
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.