Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPCR ORF4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132529
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132529 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132529 Supplier Thermo Scientific Supplier No. PA132529

Rabbit Polyclonal Antibody

PA1-32529 detects GPCR ORF4 from human samples.

The G protein-coupled receptor ORF4 is an Orphan-U GPCR with an unknown ligand. ESTs have been primarily isolated from brain, breast, cervix, eye, heart/melanocyte/uterus, lymph node, placenta, pancreas, testis, thymus, and tonsil libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPCR ORF4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR137
Gene Accession No. Q96N19
Gene Symbols GPR137
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd extracellular domain of human GPR137
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.