Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPCR v40 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132744
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
PIPA132744 25 μg
1 options

Catalog No. PIPA132744

Supplier: Thermo Scientific PA132744

Rabbit Polyclonal Antibody

PA1-32744 detects GPRv40 from human samples.

GPRv40 is an Orphan-A GPCR with no known ligand. ESTs have been isolated from human B-cell/lung/testis, brain, parathyroid, testis, and fetus/fibroblast/ovary/parathyroid/placenta libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPCR v40
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR150
Gene Accession No. Q8NGU9
Gene Alias GPRv40
Gene Symbols GPR150
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from N-terminal extracellular domain of human GPR150
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.