Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPD1L Polyclonal Antibody
GPD1L Polyclonal Antibody
Supplier: Thermo Scientific PA524216
Description
GPD1L Polyclonal Antibody for Western Blot, IHC (P), Flow

Specifications
GPD1L | |
Polyclonal | |
Unconjugated | |
GPD1L | |
2210409H23Rik; D9Ertd660e; glycerol-3-phosphate dehydrogenase 1-like; glycerol-3-phosphate dehydrogenase 1-like protein; Gpd1l; GPD1-L; Kiaa0089; RGD1560123 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23171, 333433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q3ULJ0, Q8N335 | |
GPD1L | |
KLH conjugated synthetic peptide between 44-73 amino acids from the N-terminal region of human GPD1L. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction