Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPD2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297331PCP
Description
GPD2 Polyclonal antibody specifically detects GPD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
GPD2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 1.1.5.3, GDH2, glycerol-3-phosphate dehydrogenase 2 (mitochondrial), GPDM, GPD-M, mGPDH, mitochondrial, mitochondrial glycerophosphate dehydrogenase, mtGPD | |
Produced in rabbits immunized with E. coli-derived Human GPD2 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
2820 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction