Learn More
Invitrogen™ GPD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593093
Description
Immunogen sequence: SQFAHYRRKQ MNLAYVKAAD CISEPVNREP PSREAQLLTL QNTSEFDILV IGGGATGSGC ALDAVTRGLK TALVERDDFS SGTSSRSTKL IHGGVRYLQK AIMKLDIEQY RMVKEALHER ANLLEIAPHL SAPLPIMLPV YKWWQLPYYW VGIKLYDLVA GSNCLKSSYV LSKSRALEHF PMLQKDKLVG AIVYYDGQHN DARMNLAIAL TAARYGAATA NYMEVVSLLK KTDPQTGKVR VSGARCKDVL TGQEFDVRAK CVINATGPFT DSVRKMDDKD AAAICQPSAG VHIVMPGYYS P; Positive Samples: MCF7, THP-1, SW480, Mouse skeletal muscle, Mouse liver, Mouse pancreas, Mouse brain; Cellular Location: Mitochondrion.
Mitochondrial glycerophosphate dehydrogenase (EC 1. 1. 99. 5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.
Specifications
GPD2 | |
Polyclonal | |
Unconjugated | |
Gpd2 | |
AA408484; AI448216; AU021455; AW494132; FAD-linked glycerol-3-phosphate dehydrogenase; GDH2; Gdm1; glycerol phosphate dehydrogenase 1, mitochondrial; glycerol phosphate dehydrogenase 2, mitochondrial; Glycerol-3-phosophate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2; glycerol-3-phosphate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2, mitochondrial; glycerol-3-phosphate dehydrogenase, mitochondrial; Gpd2; GPDH; GPDH-M; GPDM; GPD-M; mGPDH; mitochondrial glycerol-3-phosphate dehydrogenase; mitochondrial glycerophosphate dehydrogenase; mtGPD; mtGPDH; mtGPDH gene, promoter region and alternative transcripts; Protein TISP38; testicular tissue protein Li 76; TISP38 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14571, 25062, 2820 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35571, P43304, Q64521 | |
Gpd2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-325 of human GPD2 (NP_0003993). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.