Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPD2 Recombinant Rabbit Monoclonal Antibody (PSH0-31)
GREENER_CHOICE

Catalog No. FEMA550802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550802 Supplier Invitrogen™ Supplier No. MA550802
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: MCF-7 cell lysate, THP-1 cell lysate, U87-MG cell lysate, HeLa cell lysate, Hela cell. Predicted band size: 81 kDa Subcellular Location: Mitochondrion.

Mitochondrial glycerophosphate dehydrogenase (EC 1. 1. 99. 5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPD2
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone PSH0-31
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Gpd2
Gene Accession No. P43304
Gene Alias AA408484; AI448216; AU021455; AW494132; FAD-linked glycerol-3-phosphate dehydrogenase; GDH2; Gdm1; glycerol phosphate dehydrogenase 1, mitochondrial; glycerol phosphate dehydrogenase 2, mitochondrial; Glycerol-3-phosophate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2; glycerol-3-phosphate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2, mitochondrial; glycerol-3-phosphate dehydrogenase, mitochondrial; Gpd2; GPDH; GPDH-M; GPDM; GPD-M; mGPDH; mitochondrial glycerol-3-phosphate dehydrogenase; mitochondrial glycerophosphate dehydrogenase; mtGPD; mtGPDH; mtGPDH gene, promoter region and alternative transcripts; Protein TISP38; testicular tissue protein Li 76; TISP38
Gene Symbols Gpd2
Host Species Rabbit
Immunogen Recombinant protein within Human GPD2 aa 43-727.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.