Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPHA2 Antibody (06), FITC , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306201F
Description
GPHA2 Monoclonal antibody specifically detects GPHA2 in Human samples. It is validated for ELISASpecifications
GPHA2 | |
Monoclonal | |
FITC | |
A2, glycoprotein alpha 2, glycoprotein hormone alpha 2, glycoprotein hormone alpha-2, GPA2Thyrostimulin subunit alpha, MGC126572, Putative secreted protein Zsig51, ZSIG51cysteine knot protein | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPHA2 (Q96T91; Met1-Tyr129). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
06 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
170589 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction