Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPHA2 Antibody (064), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290250UV
Description
GPHA2 Monoclonal specifically detects GPHA2 in Human samples. It is validated for ELISA.Specifications
GPHA2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
170589 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
64 | |
ELISA | |
A2, glycoprotein alpha 2, glycoprotein hormone alpha 2, glycoprotein hormone alpha-2, GPA2Thyrostimulin subunit alpha, MGC126572, Putative secreted protein Zsig51, ZSIG51cysteine knot protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human GPHA2 (Q96T91; Met1-Tyr129). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction